SY3H3D681D89 Synthonix Corporation

3-(1-Pyrrolidylmethyl)benzoic Acid Hydrochloride

  • MDL number No MDL found
  • Cas Number : 887922-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56596.html

Safety Information

MSDS https://synthonix.com/56596.html
Availability details for
Quantity
  • Availability
  • Add to Cart