SY3H3D681D8A Synthonix Corporation

1-Phenylpiperidine-4-carboxylic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 209005-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56598.html

Safety Information

MSDS https://synthonix.com/56598.html
Availability details for
Quantity
  • Availability
  • Add to Cart