SY3H3D681D8D Synthonix Corporation

(S)-2-Phenethylpiperazine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 2055848-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56641.html

Safety Information

MSDS https://synthonix.com/56641.html
Availability details for
Quantity
  • Availability
  • Add to Cart