SY3H3D681D8E Synthonix Corporation

3-Phenyl-1H-indazole

  • MDL number No MDL found
  • Cas Number : 13097-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/56649.html

Safety Information

MSDS https://synthonix.com/56649.html
Availability details for
Quantity
  • Availability
  • Add to Cart