SY3H3D681D8F Synthonix Corporation

1-Phenyl-1H-benzo[d]imidazole

  • Cas Number : 2622-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56650.html

Safety Information

MSDS https://synthonix.com/56650.html
Availability details for
Quantity
  • Availability
  • Add to Cart