SY3H3D681D90 Synthonix Corporation

6-Phenylbenzo[d]isoxazol-3-amine

  • MDL number No MDL found
  • Cas Number : 268734-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56653.html

Safety Information

MSDS https://synthonix.com/56653.html
Availability details for
Quantity
  • Availability
  • Add to Cart