SY3H3D681D95 Synthonix Corporation

6-Isopropyl-4-oxo-4H-chromene-3-carbonitrile

  • Cas Number : 50743-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56715.html

Safety Information

MSDS https://synthonix.com/56715.html
Availability details for
Quantity
  • Availability
  • Add to Cart