SY3H3D681D97 Synthonix Corporation

2-(Phenylamino)benzamide

  • MDL number No MDL found
  • Cas Number : 1211-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56748.html

Safety Information

MSDS https://synthonix.com/56748.html
Availability details for
Quantity
  • Availability
  • Add to Cart