SY3H3D681D98 Synthonix Corporation

S-Phenyl 4-methylbenzenesulfonothioate

  • MDL number No MDL found
  • Cas Number : 3541-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56782.html

Safety Information

MSDS https://synthonix.com/56782.html
Availability details for
Quantity
  • Availability
  • Add to Cart