SY3H3D681D99 Synthonix Corporation

Phenyl p-Toluenesulfonate

  • Cas Number : 640-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin USA
Product Link https://synthonix.com/56792.html

Safety Information

MSDS https://synthonix.com/56792.html
Availability details for
Quantity
  • Availability
  • Add to Cart