SY3H3D681D9E Synthonix Corporation

N-(2-Phenoxyphenyl)methanesulfonamide

  • Cas Number : 51765-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56828.html

Safety Information

MSDS https://synthonix.com/56828.html
Availability details for
Quantity
  • Availability
  • Add to Cart