SY3H3D681DA0 Synthonix Corporation

1-(Piperidin-4-yl)-2-(trifluoromethyl)-1H-benzo[d]imidazole

  • MDL number No MDL found
  • Cas Number : 742076-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56856.html

Safety Information

MSDS https://synthonix.com/56856.html
Availability details for
Quantity
  • Availability
  • Add to Cart