SY3H3D681DA2 Synthonix Corporation

2-(2-Phenylacetamido)pentanedioic acid

  • MDL number No MDL found
  • Cas Number : 2752-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56932.html

Safety Information

MSDS https://synthonix.com/56932.html
Availability details for
Quantity
  • Availability
  • Add to Cart