SY3H3D681DA3 Synthonix Corporation

1-Phenylcyclohexanecarboxylic acid

  • MDL number No MDL found
  • Cas Number : 1135-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/56974.html

Safety Information

MSDS https://synthonix.com/56974.html
Availability details for
Quantity
  • Availability
  • Add to Cart