SY3H3D681DAA Synthonix Corporation

((S)-1-((S)-1-Phenylethyl)pyrrolidin-3-yl)methanol

  • MDL number No MDL found
  • Cas Number : 173724-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57160.html

Safety Information

MSDS https://synthonix.com/57160.html
Availability details for
Quantity
  • Availability
  • Add to Cart