SY3H3D681DAB Synthonix Corporation

2,2,4,6,7-Pentamethyl-2,3-dihydrobenzofuran-5-sulfonamide

  • MDL number No MDL found
  • Cas Number : 378230-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57172.html

Safety Information

MSDS https://synthonix.com/57172.html
Availability details for
Quantity
  • Availability
  • Add to Cart