SY3H3D681DAC Synthonix Corporation

2-(4-Isopropylphenoxy)-N,N-dimethylethanamine

  • MDL number No MDL found
  • Cas Number : 1001966-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/57228.html

Safety Information

MSDS https://synthonix.com/57228.html
Availability details for
Quantity
  • Availability
  • Add to Cart