SY3H3D681DAF Synthonix Corporation

1,10-Phenanthroline-5-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 91804-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57310.html

Safety Information

MSDS https://synthonix.com/57310.html
Availability details for
Quantity
  • Availability
  • Add to Cart