SY3H3D681DB3 Synthonix Corporation

3-Phenyl-[1,2,4]triazolo[4,3-a]pyridine-6-carboxylic acid

  • Cas Number : 936074-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57359.html

Safety Information

MSDS https://synthonix.com/57359.html
Availability details for
Quantity
  • Availability
  • Add to Cart