SY3H3D681DB5 Synthonix Corporation

Potassium (3-(benzyloxy)phenyl)trifluoroborate

  • MDL number No MDL found
  • Cas Number : 850623-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57394.html

Safety Information

MSDS https://synthonix.com/57394.html
Availability details for
Quantity
  • Availability
  • Add to Cart