SY3H3D681DB7 Synthonix Corporation

1-(2-Phenoxyphenyl)methanamine hydrochloride

  • Cas Number : 31963-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57407.html

Safety Information

MSDS https://synthonix.com/57407.html
Availability details for
Quantity
  • Availability
  • Add to Cart