SY3H3D681DBE Synthonix Corporation

(3-((2-Isopropyl-1H-imidazol-1-yl)methyl)phenyl)amine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 1211449-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57454.html

Safety Information

MSDS https://synthonix.com/57454.html
Availability details for
Quantity
  • Availability
  • Add to Cart