SY3H3D681DC0 Synthonix Corporation

5-Phenoxyisobenzofuran-1(3H)-one

  • Cas Number : 57830-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57495.html

Safety Information

MSDS https://synthonix.com/57495.html
Availability details for
Quantity
  • Availability
  • Add to Cart