SY3H3D681DC1 Synthonix Corporation

1-Phenyl-1H-indole

  • MDL number No MDL found
  • Cas Number : 16096-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/57501.html

Safety Information

MSDS https://synthonix.com/57501.html
Availability details for
Quantity
  • Availability
  • Add to Cart