SY3H3D681DC4 Synthonix Corporation

4-Phenyl-3,4-dihydro-2H-benzo[e][1,3]oxazin-2-one

  • MDL number No MDL found
  • Cas Number : 1129278-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57511.html

Safety Information

MSDS https://synthonix.com/57511.html
Availability details for
Quantity
  • Availability
  • Add to Cart