SY3H3D681DC6 Synthonix Corporation

2-Phenyl-1H-indol-5-amine

  • MDL number No MDL found
  • Cas Number : 6855-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57545.html

Safety Information

MSDS https://synthonix.com/57545.html
Availability details for
Quantity
  • Availability
  • Add to Cart