SY3H3D681DC9 Synthonix Corporation

1-Phenoxy-4-vinylbenzene

  • MDL number No MDL found
  • Cas Number : 4973-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57565.html

Safety Information

MSDS https://synthonix.com/57565.html
Availability details for
Quantity
  • Availability
  • Add to Cart