SY3H3D681DCF Synthonix Corporation

1-(3-(Phenylamino)phenyl)ethanone

  • Cas Number : 23699-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57607.html

Safety Information

MSDS https://synthonix.com/57607.html
Availability details for
Quantity
  • Availability
  • Add to Cart