SY3H3D681DD4 Synthonix Corporation

3-Phenyl-2-tosyl-1,2-oxaziridine

  • Cas Number : 63160-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57623.html

Safety Information

MSDS https://synthonix.com/57623.html
Availability details for
Quantity
  • Availability
  • Add to Cart