SY3H3D681DD6 Synthonix Corporation

4-Propyl-1-naphthoic acid

  • MDL number No MDL found
  • Cas Number : 107777-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57652.html

Safety Information

MSDS https://synthonix.com/57652.html
Availability details for
Quantity
  • Availability
  • Add to Cart