SY3H3D6821C6 Synthonix Corporation

4-Phenyl-2,8-diazaspiro[4.5]decan-3-one hydrochloride

  • Cas Number : 2044705-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58088.html

Safety Information

MSDS https://synthonix.com/58088.html
Availability details for
Quantity
  • Availability
  • Add to Cart