SY3H3D6821C7 Synthonix Corporation

Sodium 2-hydroxy-2-(4-hydroxyphenyl)acetate

  • MDL number No MDL found
  • Cas Number : 69322-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/49483.html

Safety Information

MSDS https://synthonix.com/49483.html
Availability details for
Quantity
  • Availability
  • Add to Cart