SY3H3D6821C9 Synthonix Corporation

1-(Thiazolidin-2-yl)ethanone hydrochloride

  • MDL number No MDL found
  • Cas Number : 1993017-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41381.html

Safety Information

MSDS https://synthonix.com/41381.html
Availability details for
Quantity
  • Availability
  • Add to Cart