SY3H3D6821CA Synthonix Corporation

4,5,6,7-Tetrahydro-1H-benzo[d]imidazole-5-carboxylic acid

  • MDL number MFCD03839866
  • Cas Number : 26751-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46687.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46687.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart