SY3H3D6821CC Synthonix Corporation

N-[2-Isopropylthiazol-4-ylmethyl(methyl)carbamoyl]-L-valine

  • MDL number No MDL found
  • Cas Number : 154212-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57967.html

Safety Information

MSDS https://synthonix.com/57967.html
Availability details for
Quantity
  • Availability
  • Add to Cart