SY3H3D6821CD Synthonix Corporation

(2R,5R)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 5-hydroxy-1,3-oxathiolane-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 147126-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57983.html

Safety Information

MSDS https://synthonix.com/57983.html
Availability details for
Quantity
  • Availability
  • Add to Cart