SY3H3D6821D0 Synthonix Corporation

1,10-Phenanthroline-4,7-dicarboxylic acid

  • MDL number No MDL found
  • Cas Number : 31301-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58033.html

Safety Information

MSDS https://synthonix.com/58033.html
Availability details for
Quantity
  • Availability
  • Add to Cart