SY3H3D6821D1 Synthonix Corporation

1-(Phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine-6-carbonitrile

  • Cas Number : 1227270-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58052.html

Safety Information

MSDS https://synthonix.com/58052.html
Availability details for
Quantity
  • Availability
  • Add to Cart