SY3H3D6821D6 Synthonix Corporation

3-(Piperidin-4-ylmethyl)-1H-indole

  • MDL number No MDL found
  • Cas Number : 3515-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57768.html

Safety Information

MSDS https://synthonix.com/57768.html
Availability details for
Quantity
  • Availability
  • Add to Cart