SY3H3D6821DF Synthonix Corporation

5-Phenyl-1H-indole-3-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 141835-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58140.html

Safety Information

MSDS https://synthonix.com/58140.html
Availability details for
Quantity
  • Availability
  • Add to Cart