SY3H3D6821E1 Synthonix Corporation

3-Phenyl-2,3-dihydro-1H-inden-1-one

  • MDL number No MDL found
  • Cas Number : 16618-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/58164.html

Safety Information

MSDS https://synthonix.com/58164.html
Availability details for
Quantity
  • Availability
  • Add to Cart