SY3H3D6821E2 Synthonix Corporation

4-Phenylcinnamic acid

  • Cas Number : 13026-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58165.html

Safety Information

MSDS https://synthonix.com/58165.html
Availability details for
Quantity
  • Availability
  • Add to Cart