SY3H3D6821E8 Synthonix Corporation

4-Propyl-1,1'-biphenyl

  • MDL number MFCD00102114
  • Cas Number : 10289-45-9
  • Molecular Weight : 196.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Clear liquid - Slightly cloudy
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/58267.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/58267.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart