SY3H3D6821EB Synthonix Corporation

1-(5-Phenylpyridin-3-yl)piperazine

  • Cas Number : 223794-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58288.html

Safety Information

MSDS https://synthonix.com/58288.html
Availability details for
Quantity
  • Availability
  • Add to Cart