SY3H3D6821ED Synthonix Corporation

5-(4-Isopropoxy-3-methylphenyl)-1H-pyrrole-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1517011-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58301.html

Safety Information

MSDS https://synthonix.com/58301.html
Availability details for
Quantity
  • Availability
  • Add to Cart