SY3H3D6821F3 Synthonix Corporation

4-(4-Isopropoxyphenyl)cyclohexanone

  • MDL number No MDL found
  • Cas Number : 365553-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58384.html

Safety Information

MSDS https://synthonix.com/58384.html
Availability details for
Quantity
  • Availability
  • Add to Cart