SY3H3D6821F5 Synthonix Corporation

3-Isopropyl-2-(isopropylimino)-5-phenyl-1,3,5-thiadiazinan-4-one

  • MDL number No MDL found
  • Cas Number : 69327-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/58394.html

Safety Information

MSDS https://synthonix.com/58394.html
Availability details for
Quantity
  • Availability
  • Add to Cart