SY3H3D6821F7 Synthonix Corporation

3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-amine

  • MDL number No MDL found
  • Cas Number : 116233-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58447.html

Safety Information

MSDS https://synthonix.com/58447.html
Availability details for
Quantity
  • Availability
  • Add to Cart