SY3H3D6821F8 Synthonix Corporation

(4-(4-Propylcyclohexyl)phenyl)boronic acid

  • Cas Number : 156837-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58465.html

Safety Information

MSDS https://synthonix.com/58465.html
Availability details for
Quantity
  • Availability
  • Add to Cart