SY3H3D6821F9 Synthonix Corporation

(4-(trans-4-propylcyclohexyl)phenyl)boronic acid

  • Cas Number : 146862-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58466.html

Safety Information

MSDS https://synthonix.com/58466.html
Availability details for
Quantity
  • Availability
  • Add to Cart